Assay Details
| Gene Symbol: acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212620.2 | NP_997785.2 | 6 - 7 | 713 | 68 | |
| GenBank mRNA | AY398305.1 | - | 6 - 7 | 703 | 68 | |
| BC075896.1 | - | 6 - 7 | 681 | 68 | ||
More Information
Set Membership: |
|

acta2
Genomic Map