Assay Details
| Gene Symbol: fbxw11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213504.2 | NP_998669.2 | 10 - 11 | 1377 | 91 | |
| XM_005155294.4 | XP_005155351.1 | 10 - 11 | 1388 | 91 | ||
| GenBank mRNA | BC056809.1 | - | 10 - 11 | 1379 | 91 | |
More Information
Set Membership: |
|

fbxw11b
Genomic Map