Assay Details
| Gene Symbol: fbxw11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213504.2 | NP_998669.2 | 12 - 13 | 1629 | 77 | |
| XM_005155294.4 | XP_005155351.1 | 12 - 13 | 77 | |||
| GenBank mRNA | BC056809.1 | - | 12 - 13 | 1631 | 77 | |
More Information
Set Membership: |
|

fbxw11b
Genomic Map