Assay Details
| Gene Symbol: fbxw11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213504.2 | NP_998669.2 | 2 - 3 | 126 | 85 | |
| GenBank mRNA | BC056809.1 | - | 2 - 3 | 128 | 85 | |
More Information
Set Membership: |
|

fbxw11b
Genomic Map