Assay Details
| Gene Symbol: lsm14ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214772.2 | NP_999937.2 | 7 - 8 | 1216 | 74 | |
| XM_005158973.3 | XP_005159030.1 | 8 - 9 | 1315 | 74 | ||
| XM_005158974.3 | XP_005159031.1 | 8 - 9 | 1315 | 74 | ||
| GenBank mRNA | AY577000.1 | - | 7 - 8 | 1193 | 74 | |
| BC047185.1 | - | 7 - 8 | 1213 | 74 | ||
| BC065685.1 | - | 7 - 8 | 1181 | 74 | ||
| BC165583.1 | - | 7 - 8 | 1075 | 74 | ||
More Information
Set Membership: |
|

lsm14ab
Genomic Map