Assay Details
| Gene Symbol: puf60b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002121.1 | NP_001002121.1 | 10 - 11 | 1320 | 99 | |
| XM_009303395.3 | XP_009301670.1 | 11 - 12 | 1509 | 99 | ||
| GenBank mRNA | BC071467.1 | - | 10 - 11 | 1320 | 99 | |
More Information
Set Membership: |
|

puf60b
Genomic Map