Assay Details
| Gene Symbol: puf60b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002121.1 | NP_001002121.1 | 4 - 5 | 285 | 119 | |
| GenBank mRNA | BC071467.1 | - | 4 - 5 | 285 | 119 | |
More Information
Set Membership: |
|

puf60b
Genomic Map