Assay Details
| Gene Symbol: puf60b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002121.1 | NP_001002121.1 | 5 - 6 | 440 | 72 | |
| XM_009303395.3 | XP_009301670.1 | 6 - 7 | 629 | 72 | ||
| GenBank mRNA | BC071467.1 | - | 5 - 6 | 440 | 72 | |
More Information
Set Membership: |
|

puf60b
Genomic Map