Assay Details
| Gene Symbol: polr1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003888.1 | NP_001003888.1 | 2 - 3 | 151 | 76 | |
| GenBank mRNA | AY648775.1 | - | 2 - 3 | 151 | 76 | |
| BC115138.1 | - | 2 - 3 | 151 | 76 | ||
More Information
Set Membership: |
|

polr1d
Genomic Map