Assay Details
| Gene Symbol: tnni1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008613.1 | NP_001008613.1 | 6 - 7 | 380 | 71 | |
| GenBank mRNA | BC086814.1 | - | 6 - 7 | 380 | 71 | |
| BC164964.1 | - | 3 - 4 | 305 | 71 | ||
More Information
Set Membership: |
|

tnni1b
Genomic Map