Assay Details
| Gene Symbol: fgf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012246.2 | NP_001012246.2 | 1 - 2 | 292 | 126 | |
| GenBank mRNA | AB183515.1 | - | 1 - 2 | 196 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001012246.2 transcripts.
Set Membership: |
|

fgf19
Genomic Map