Assay Details
| Gene Symbol: sema3fa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014822.1 | NP_001014822.1 | 10 - 11 | 1099 | 82 | |
| XM_005166039.3 | XP_005166096.1 | 11 - 12 | 82 | |||
| XM_005166040.3 | XP_005166097.1 | 10 - 11 | 82 | |||
| GenBank mRNA | AY766118.1 | - | 10 - 11 | 1099 | 82 | |
| BC163764.1 | - | 10 - 11 | 82 | |||
| BC163771.1 | - | 10 - 11 | 82 | |||
| CU458859.1 | - | 10 - 11 | 1114 | 82 | ||
More Information
Set Membership: |
|

sema3fa
Genomic Map