Assay Details
| Gene Symbol: sema3fa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014822.1 | NP_001014822.1 | 13 - 14 | 1561 | 91 | |
| XM_005166039.3 | XP_005166096.1 | 14 - 15 | 2106 | 91 | ||
| XM_005166040.3 | XP_005166097.1 | 13 - 14 | 2016 | 91 | ||
| GenBank mRNA | AY766118.1 | - | 13 - 14 | 1561 | 91 | |
| BC163764.1 | - | 13 - 14 | 1617 | 91 | ||
| BC163771.1 | - | 13 - 14 | 1617 | 91 | ||
| CU458859.1 | - | 13 - 14 | 1576 | 91 | ||
More Information
Set Membership: |
|

sema3fa
Genomic Map