Assay Details
| Gene Symbol: sema3fa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014822.1 | NP_001014822.1 | 15 - 16 | 1767 | 64 | |
| XM_005166039.3 | XP_005166096.1 | 16 - 17 | 2312 | 64 | ||
| XM_005166040.3 | XP_005166097.1 | 15 - 16 | 2222 | 64 | ||
| GenBank mRNA | AY766118.1 | - | 15 - 16 | 1767 | 64 | |
| BC163764.1 | - | 15 - 16 | 1823 | 64 | ||
| BC163771.1 | - | 15 - 16 | 1823 | 64 | ||
| CU458859.1 | - | 15 - 16 | 1782 | 64 | ||
More Information
Set Membership: |
|

sema3fa
Genomic Map