Assay Details
| Gene Symbol: sema3fa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014822.1 | NP_001014822.1 | 9 - 10 | 1033 | 85 | |
| XM_005166039.3 | XP_005166096.1 | 10 - 11 | 1578 | 85 | ||
| XM_005166040.3 | XP_005166097.1 | 9 - 10 | 1488 | 85 | ||
| GenBank mRNA | AY766118.1 | - | 9 - 10 | 1033 | 85 | |
| BC163764.1 | - | 9 - 10 | 1089 | 85 | ||
| BC163771.1 | - | 9 - 10 | 1089 | 85 | ||
| CU458859.1 | - | 9 - 10 | 1048 | 85 | ||
More Information
Set Membership: |
|

sema3fa
Genomic Map