Assay Details
| Gene Symbol: fabp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020558.1 | NP_001018394.1 | 2 - 3 | 333 | 80 | |
| GenBank mRNA | BC095142.1 | - | 2 - 3 | 333 | 80 | |
| BC165498.1 | - | 2 - 3 | 289 | 80 | ||
| DQ266470.1 | - | 2 - 3 | 255 | 80 | ||
More Information
Set Membership: |
|

fabp4b
Genomic Map