Assay Details
| Gene Symbol: tent2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020600.1 | NP_001018436.1 | 9 - 10 | 994 | 88 | |
| XM_021476157.1 | XP_021331832.1 | 9 - 10 | 88 | |||
| GenBank mRNA | BC095312.1 | - | 9 - 10 | 994 | 88 | |
More Information
Set Membership: |
|

tent2
Genomic Map