Assay Details
| Gene Symbol: ak8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020644.1 | NP_001018480.1 | 9 - 10 | 891 | 63 | |
| GenBank mRNA | BC095649.1 | - | 9 - 10 | 891 | 63 | |
More Information
Set Membership: |
|

ak8
Genomic Map