Assay Details
| Gene Symbol: prelid1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020679.1 | NP_001018515.1 | 4 - 5 | 511 | 83 | |
| XM_017353568.2 | XP_017209057.1 | 4 - 5 | 505 | 83 | ||
| GenBank mRNA | BC095754.1 | - | 4 - 5 | 511 | 83 | |
More Information
Set Membership: |
|

prelid1b
Genomic Map