Assay Details
| Gene Symbol: prelid1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020679.1 | NP_001018515.1 | 5 - 6 | 594 | 121 | |
| GenBank mRNA | BC095754.1 | - | 5 - 6 | 594 | 121 | |
More Information
Set Membership: |
|

prelid1b
Genomic Map