Assay Details
| Gene Symbol: tdg.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020751.1 | NP_001018587.1 | 2 - 3 | 129 | 87 | |
| GenBank mRNA | BC095340.1 | - | 2 - 3 | 129 | 87 | |
| BC165885.1 | - | 1 - 2 | 93 | 87 | ||
More Information
Set Membership: |
|

tdg.1
Genomic Map