Assay Details
| Gene Symbol: tdg.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020751.1 | NP_001018587.1 | 3 - 4 | 302 | 101 | |
| XM_005163014.4 | XP_005163071.1 | 2 - 3 | 303 | 101 | ||
| GenBank mRNA | BC095340.1 | - | 3 - 4 | 302 | 101 | |
| BC165885.1 | - | 2 - 3 | 266 | 101 | ||
More Information
Set Membership: |
|

tdg.1
Genomic Map