Assay Details
| Gene Symbol: ezrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030285.2 | NP_001025456.1 | 8 - 9 | 901 | 114 | |
| XM_005160437.4 | XP_005160494.1 | 8 - 9 | 892 | 114 | ||
| GenBank mRNA | BC078311.1 | - | 8 - 9 | 998 | 114 | |
| BC139506.1 | - | 8 - 9 | 958 | 114 | ||
More Information
Set Membership: |
|

ezrb
Genomic Map