Assay Details
| Gene Symbol: ezrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030285.2 | NP_001025456.1 | 10 - 11 | 1194 | 64 | |
| XM_005160437.4 | XP_005160494.1 | 10 - 11 | 1185 | 64 | ||
| GenBank mRNA | BC139506.1 | - | 10 - 11 | 1251 | 64 | |
More Information
Set Membership: |
|

ezrb
Genomic Map