Assay Details
| Gene Symbol: ppfibp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044970.1 | NP_001038435.1 | 23 - 24 | 2695 | 101 | |
| XM_009293503.3 | XP_009291778.1 | 23 - 24 | 2964 | 101 | ||
| XM_009293504.3 | XP_009291779.1 | 22 - 23 | 2931 | 101 | ||
| XM_021467668.1 | XP_021323343.1 | 24 - 25 | 3057 | 101 | ||
| XM_021467669.1 | XP_021323344.1 | 23 - 24 | 3024 | 101 | ||
| XM_021467670.1 | XP_021323345.1 | 23 - 24 | 2997 | 101 | ||
| GenBank mRNA | BC055152.1 | - | 8 - 9 | 879 | 101 | |
| BC139625.1 | - | 6 - 7 | 611 | 101 | ||
| BC171534.1 | - | 22 - 23 | 2413 | 101 | ||
More Information
Set Membership: |
|

ppfibp1b
Genomic Map