Assay Details
| Gene Symbol: alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045354.1 | NP_001038819.1 | 7 - 8 | 741 | 79 | |
| GenBank mRNA | BC122259.1 | - | 7 - 8 | 741 | 79 | |
| BC164125.1 | - | 7 - 8 | 655 | 79 | ||
More Information
Set Membership: |
|

alg5
Genomic Map