Assay Details
| Gene Symbol: grsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045852.1 | NP_001039317.1 | 2 - 3 | 379 | 100 | |
| XM_005165334.2 | XP_005165391.1 | 2 - 3 | 534 | 100 | ||
| GenBank mRNA | BC122226.1 | - | 2 - 3 | 379 | 100 | |
| BC164250.1 | - | 2 - 3 | 210 | 100 | ||
More Information
Set Membership: |
|

grsf1
Genomic Map