Assay Details
| Gene Symbol: setdb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077277.3 | NP_001070745.3 | 7 - 8 | 1227 | 125 | |
| XM_005158013.4 | XP_005158070.2 | 9 - 10 | 1438 | 125 | ||
| XM_005158015.4 | XP_005158072.2 | 9 - 10 | 1438 | 125 | ||
| XM_005158016.4 | XP_005158073.2 | 7 - 8 | 1237 | 125 | ||
| GenBank mRNA | BC124601.1 | - | 7 - 8 | 1170 | 125 | |
| BC165176.1 | - | 6 - 7 | 1014 | 125 | ||
More Information
Set Membership: |
|

setdb1b
Genomic Map