Assay Details
| Gene Symbol: ino80db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080588.1 | NP_001074057.1 | 6 - 7 | 1781 | 78 | |
| XM_009302237.3 | XP_009300512.1 | 6 - 7 | 2658 | 78 | ||
| XM_009302238.3 | XP_009300513.1 | 6 - 7 | 2673 | 78 | ||
| GenBank mRNA | BC129037.1 | - | 6 - 7 | 1781 | 78 | |
More Information
Set Membership: |
|

ino80db
Genomic Map