Assay Details
| Gene Symbol: morc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089366.1 | NP_001082835.1 | 15 - 16 | 1674 | 64 | |
| GenBank mRNA | BC135000.1 | - | 15 - 16 | 1674 | 64 | |
More Information
Set Membership: |
|

morc3a
Genomic Map