Assay Details
| Gene Symbol: morc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089366.1 | NP_001082835.1 | 1 - 2 | 70 | 65 | |
| GenBank mRNA | BC095325.1 | - | 1 - 2 | 110 | 65 | |
| BC135000.1 | - | 1 - 2 | 70 | 65 | ||
More Information
Set Membership: |
|

morc3a
Genomic Map