Assay Details
| Gene Symbol: matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099740.2 | NP_001093210.1 | 2 - 3 | 487 | 56 | |
| XM_021468031.1 | XP_021323706.1 | 2 - 3 | 505 | 56 | ||
| GenBank mRNA | AJ629191.1 | - | 2 - 3 | 460 | 56 | |
| AJ697718.1 | - | 2 - 3 | 460 | 56 | ||
| BC045465.1 | - | 2 - 3 | 483 | 56 | ||
More Information
Set Membership: |
|

matn1
Genomic Map