Assay Details
| Gene Symbol: tbx3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101670.2 | NP_001095140.2 | 3 - 4 | 1247 | 99 | |
| GenBank mRNA | BC133068.1 | - | 3 - 4 | 1238 | 99 | |
More Information
Set Membership: |
|

tbx3a
Genomic Map