Assay Details
| Gene Symbol: tbx3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101670.2 | NP_001095140.2 | 4 - 5 | 1327 | 125 | |
| GenBank mRNA | BC133068.1 | - | 4 - 5 | 1318 | 125 | |
More Information
Set Membership: |
|

tbx3a
Genomic Map