Assay Details
| Gene Symbol: zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109824.1 | NP_001103294.1 | 7 - 8 | 1634 | 66 | |
| XM_005157926.4 | XP_005157983.1 | 7 - 8 | 66 | |||
| XM_021466716.1 | XP_021322391.1 | 7 - 8 | 66 | |||
| XM_021466717.1 | XP_021322392.1 | 5 - 6 | 66 | |||
| GenBank mRNA | BC124312.1 | - | 7 - 8 | 1499 | 66 | |
| BC151921.1 | - | 7 - 8 | 1634 | 66 | ||
More Information
Set Membership: |
|

zyx
Genomic Map