Assay Details
| Gene Symbol: pcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131404.2 | NP_571479.2 | 3 - 4 | 652 | 94 | |
| GenBank mRNA | AF140608.1 | - | 3 - 4 | 429 | 94 | |
| BC049535.1 | - | 3 - 4 | 592 | 94 | ||
| BC064299.1 | - | 3 - 4 | 570 | 94 | ||
| EH446205.1 | - | 3 - 4 | 681 | 94 | ||
More Information
Set Membership: |
|

pcna
Genomic Map