Assay Details
| Gene Symbol: scn8aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131628.1 | NP_571703.1 | 12 - 13 | 2188 | 102 | |
| XM_009296874.3 | XP_009295149.1 | 12 - 13 | 2198 | 102 | ||
| GenBank mRNA | AF297658.1 | - | 12 - 13 | 2188 | 102 | |
More Information
Set Membership: |
|

scn8aa
Genomic Map