Assay Details
| Gene Symbol: scn8aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131628.1 | NP_571703.1 | 14 - 15 | 2560 | 68 | |
| XM_005162268.4 | XP_005162325.1 | 14 - 15 | 2604 | 68 | ||
| XM_009296871.3 | XP_009295146.1 | 14 - 15 | 2604 | 68 | ||
| XM_009296874.3 | XP_009295149.1 | 14 - 15 | 2570 | 68 | ||
| XM_021469841.1 | XP_021325516.1 | 14 - 15 | 2749 | 68 | ||
| XM_021469842.1 | XP_021325517.1 | 14 - 15 | 2618 | 68 | ||
| XM_021469843.1 | XP_021325518.1 | 14 - 15 | 2617 | 68 | ||
| GenBank mRNA | AF297658.1 | - | 14 - 15 | 2560 | 68 | |
| BC163773.1 | - | 14 - 15 | 2414 | 68 | ||
More Information
Set Membership: |
|

scn8aa
Genomic Map