Assay Details
| Gene Symbol: dcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131697.1 | NP_571772.1 | 7 - 8 | 1005 | 64 | |
| GenBank mRNA | AB906691.1 | - | 7 - 8 | 1005 | 64 | |
| AB906692.1 | - | 7 - 8 | 1005 | 64 | ||
| AB906693.1 | - | 7 - 8 | 1005 | 64 | ||
| BC044206.1 | - | 7 - 8 | 1005 | 64 | ||
| CU468786.1 | - | 6 - 7 | 921 | 64 | ||
More Information
Set Membership: |
|

dcn
Genomic Map