Assay Details
| Gene Symbol: cebpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131886.1 | NP_571961.1 | 2 - 3 | 336 | 107 | |
| GenBank mRNA | AF306856.1 | - | 2 - 3 | 336 | 107 | |
| BC049400.1 | - | 2 - 3 | 346 | 107 | ||
| BC071296.1 | - | 2 - 3 | 361 | 107 | ||
More Information
Set Membership: |
|

cebpg
Genomic Map