Assay Details
| Gene Symbol: p2rx4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153653.1 | NP_705939.1 | 9 - 10 | 1006 | 93 | |
| XM_009295279.3 | XP_009293554.1 | 8 - 9 | 784 | 93 | ||
| GenBank mRNA | AF317643.1 | - | 9 - 10 | 1006 | 93 | |
| BC042317.1 | - | 9 - 10 | 1166 | 93 | ||
| BC066495.1 | - | 9 - 10 | 995 | 93 | ||
| BC164717.1 | - | 9 - 10 | 1049 | 93 | ||
More Information
Set Membership: |
|

p2rx4a
Genomic Map