Assay Details
| Gene Symbol: hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200019.1 | NP_956313.1 | 11 - 12 | 1095 | 67 | |
| GenBank mRNA | BC046041.1 | - | 11 - 12 | 1095 | 67 | |
More Information
Set Membership: |
|

hadhb
Genomic Map