Assay Details
| Gene Symbol: stimate | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200467.1 | NP_956761.1 | 5 - 6 | 782 | 72 | |
| GenBank mRNA | BC055172.1 | - | 5 - 6 | 782 | 72 | |
More Information
Set Membership: |
|

stimate
Genomic Map