Assay Details
| Gene Symbol: creld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200523.1 | NP_956817.1 | 4 - 5 | 574 | 60 | |
| GenBank mRNA | BC055626.1 | - | 4 - 5 | 574 | 60 | |
| KM655615.1 | - | 4 - 5 | 409 | 60 | ||
More Information
Set Membership: |
|

creld2
Genomic Map