Assay Details
| Gene Symbol: rpe65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200751.1 | NP_957045.1 | 13 - 14 | 1480 | 80 | |
| GenBank mRNA | AY646886.1 | - | 12 - 13 | 1447 | 80 | |
| BC059559.1 | - | 13 - 14 | 1480 | 80 | ||
More Information
Set Membership: |
|

rpe65a
Genomic Map