Assay Details
| Gene Symbol: rpe65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200751.1 | NP_957045.1 | 6 - 7 | 679 | 69 | |
| GenBank mRNA | AY646886.1 | - | 5 - 6 | 646 | 69 | |
| BC059559.1 | - | 6 - 7 | 679 | 69 | ||
More Information
Set Membership: |
|

rpe65a
Genomic Map