Assay Details
| Gene Symbol: hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201496.1 | NP_958904.1 | 2 - 3 | 598 | 77 | |
| GenBank mRNA | BC044475.1 | - | 2 - 3 | 598 | 77 | |
| BC164149.1 | - | 2 - 3 | 432 | 77 | ||
More Information
Set Membership: |
|

hif1an
Genomic Map