Assay Details
| Gene Symbol: eef1akmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207087.1 | NP_996970.1 | 3 - 4 | 277 | 58 | |
| XM_005167551.4 | XP_005167608.1 | 3 - 4 | 58 | |||
| GenBank mRNA | BC066508.1 | - | 3 - 4 | 277 | 58 | |
More Information
Set Membership: |
|

eef1akmt1
Genomic Map