Assay Details
| Gene Symbol: tmem263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213141.1 | NP_998306.1 | 2 - 3 | 280 | 116 | |
| GenBank mRNA | AY398380.1 | - | 2 - 3 | 290 | 116 | |
| BC053251.1 | - | 2 - 3 | 280 | 116 | ||
More Information
Set Membership: |
|

tmem263
Genomic Map