Assay Details
| Gene Symbol: wipf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002165.1 | NP_001002165.1 | 4 - 5 | 428 | 70 | |
| XM_005173756.4 | XP_005173813.1 | 4 - 5 | 483 | 70 | ||
| GenBank mRNA | BC071180.1 | - | 4 - 5 | 428 | 70 | |
More Information
Set Membership: |
|

wipf2b
Genomic Map